Replacement Parts for C20 Karl Fischer™ Autotitrator
Actual product may vary
Mettler Toledo
Replacement Parts for C20 Karl Fischer™ Autotitrator
Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator
Have Questions?
Catalog NumberType
FIS01-910-744Septa
Catalog number FIS01-910-744
Price (SGD)
430.00
Each
Type:
Septa
Price (SGD)
430.00
Each
Coulometric Cells
  • Coulometer cells with or without diaphragm
  • Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
  • Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Coulometric Reagent Exchange Set
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents
Specifications
TypeSepta
For Use With (Equipment)C20 Coulometric Titrator
Unit SizeEach