Aluminum Weigh Boats for Micro and Ultramicro Series
Actual product may vary
Mettler Toledo
Aluminum Weigh Boats for Micro and Ultramicro Series
For discontinued Micro (MT5) and Ultramicro (UMT2) Series balances
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Weight (Metric)Width (Metric)Height (Metric)
FIS01-910-713 mm6 mg12.7 mm4 mm
FIS01-910-726.3 mm70 mg22 mm7 mm
Catalog number FIS01-910-71
Price (SGD)
1,341.00
Pack
Length (Metric):
3 mm
Weight (Metric):
6 mg
Width (Metric):
12.7 mm
Height (Metric):
4 mm
Price (SGD)
1,341.00
Pack
For discontinued Micro (MT5) and Ultramicro (UMT2) Series balances
Sturdy yet lightweight boats for sample handling.
Specifications
AutoclavableAutoclavable
Bottom ShapeFlat
ColorSilver
DisposableReusable
MaterialAluminum
ShapeRectangular
SterilityNon-sterile
TypeWeighing Boat
For Use With (Equipment)Discontinued Micro (MT5) and Ultramicro (UMT2) Series Balances
Dimensions (L x W x H)0.12 x 0.5 x 0.18 in. (3 x 12.7 x 4.5 mm)
Length (English)0.12 in.
Length (Metric)3 mm
Width (English)0.5 in.
Width (Metric)12.7 mm
Height (Metric)4 mm
Height (English)0.18 in.
Weight (Metric)6 mg
Unit SizePack