Zipper Seal Sample Bags
Actual product may vary
Fisherbrand
Zipper Seal Sample Bags
Zipper-locking feature securely seals bags
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Thickness (English)Material
FIS01-816-1A3 x 2 in. (7 x 5cm)2 milLDPE
FIS01-816-1B5 x 3 in. (13 x 7cm)2 milLDPE
FIS01-816C18 x 9 in. (46 x 23cm)4 milPolyethylene
FIS01-816-1C6 x 4 in. (15 x 10cm)2 milPolyethylene
FIS01-816-1D8 x 5 in. (20 x 13cm)2 milPolyethylene
FIS01-816A6 x 4 in. (15.2 x 10.16cm)4 milLDPE
FIS01-816E13 x 9 in., (33 x 23cm)4 milLDPE
FIS01-816-1E12 x 9 in. (30 x 23cm)2 milPolyethylene
FIS01-816D8 x 5 in. (20 x 13cm)4 milLDPE
FIS01-816F26 x 9 in. (66 x 23cm)4 milPolyethylene
Catalog number FIS01-816-1A
Price (SGD)
16.00
Case
Dimensions (L x W):
3 x 2 in. (7 x 5cm)
Thickness (English):
2 mil
Material:
LDPE
Price (SGD)
16.00
Case
Zipper-locking feature securely seals bags
  • Durable, transparent polyethylene
  • Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
  • Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
AutoclavableNot Autoclavable
Closure TypeZip Seal
ColorClear
MaterialLDPE
No. per Case1000
No. per Pack100
Thickness (English)2 mil
Dimensions (L x W)3 x 2 in. (7 x 5cm)
Length (English)3 in.
Width (English)2 in.
Capacity (English)5 oz.
Length (Metric)7 cm
Width (Metric)5 cm
Unit SizeCase