Autoclavable HD Biohazard Waste Bags
Autoclavable HD Biohazard Waste Bags
Actual product may vary
Autoclavable HD Biohazard Waste Bags
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)
FIS01-815C35 x 24 in.
FIS01-815A24 x 12 in.
Catalog number FIS01-815C
Price (SGD)
249.00
CS
Add to cart
Dimensions (L x W):
35 x 24 in.
Price (SGD)
249.00
CS
Add to cart
Impact- and tear-resistant autoclave bags for waste sterilization. Fisherbrand™ Orange Autoclave Bags without Sterilization Indicator are constructed of high-density polyethylene.
  • Impact- and tear-resistant.
  • Bright orange color.
  • High-density polyethylene construction
Specifications
Certifications/ComplianceASTM
Closure TypeNone
ColorOrange with Black Print
Dimensions (L x W)35 x 24 in.
Length (Metric)88.9 cm
Width (Metric)61 cm
AutoclavableAutoclavable
IncludesPrinted in English
Width (English)24 in.
Length (English)35 in.
Thickness38 micron
MaterialHDPE
Temperature (Metric)127°C
Unit SizeCS