Swing Samplers
Actual product may vary
Swing Samplers
Swing Samplers
Nasco
Swing Samplers
Specially designed for collecting samples from a horizontal flowing stream.
Have Questions?
Catalog NumberLength (English)
FIS01-812-846 to 12 ft. (extendable)
Catalog number FIS01-812-84
Price (SGD)
544.00
Each
Length (English):
6 to 12 ft. (extendable)
Price (SGD)
544.00
Each
Specially designed for collecting samples from a horizontal flowing stream
  • The end of the sampler swings, allowing collection from different angles, including 90°
  • The 6 ft. (1.83 m) adjustable fiberglass pole extends to 12 ft. (3.66 m) providing a long length for any sampling situation.
  • Unit comes with a 960 mL polyethylene bottle and the cover has a vinyl liner for leak-proof protection. The bottle is attached to a polyurethane holder and is held in place with a plastic snapper ring that has an adjustable locking device.
Specifications
Product TypeSwing Arm Sampler
MaterialFiberglass
AutoclavableNot Autoclavable
Includes960 mL polyethylene bottle (attaches to pole with plastic snapper ring), cover with vinyl liner
DisposableReusable
GraduatedYes
SterilityNon-sterile
For Use With (Application)Allowing collection of samples from different angles
Length (English)6 to 12 ft. (extendable)
Length (Metric)1.8 to 3.7 m
Unit SizeEach