Standard Sample Bags
Actual product may vary
Nasco
Standard Sample Bags
Features puncture proof tabs for protection from damage due to wire-end protrusion. Easy-to-close bags are ideal for sampling and product analysis. Available in plain or write-on styles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Thickness (English)Capacity (English)Product Type
FIS01-812-78B30 x 19cm4 mil55 oz.Sample Bag with Labeling Area
FIS01-812-12018 x 9.5 cm3 mil7 oz.Sample Bag
FIS01-812-5N19 x 13 cm3 mil13 oz.Sample Bag
FIS01-812-5B23 x 11.5 cm2.5 mil18 oz.Sample Bag
FIS01-812-5C23 x 15 cm3 mil24 oz.Sample Bag
FIS01-812-338 x 19 cm3 mil69 oz.Sample Bag
FIS01-812-6A12.5 x 7.5 cm2.25 mil2 oz.Sample Bag with Labeling Area
FIS01-812-6B18.5 x 7.5cm2.25 mil4 oz.Sample Bag with Labeling Area
FIS01-812-6C23 x 11.5cm2.5 mil18 oz.Sample Bag with Labeling Area
FKS1177196323 x 11.5cm2.5 mil18 oz.Sample Bag with Labeling Area
FIS01-812-1A23 x 15cm3 mil24 oz.Sample Bag with Labeling Area
Catalog number FIS01-812-78B
Price (SGD)
746.00
Pack
Dimensions (L x W):
30 x 19cm
Thickness (English):
4 mil
Capacity (English):
55 oz.
Product Type:
Sample Bag with Labeling Area
Price (SGD)
746.00
Pack
Features puncture proof tabs for protection from damage due to wire-end protrusion. Easy-to-close bags are ideal for sampling and product analysis. Available in plain or write-on styles.
  • Transparent polyethylene bags
  • Whirling or folding thebag over three times and securing with the closure tabs creates a leak-proof sampling container
  • Gas sterilized, with documentation available
  • Pull tabs help prevent contamination from finger contact when opening
  • Write-on bags have area that accepts ballpoint pen or pencil
  • Bags can be frozen to any temperature; including liquid nitrogen--careful handling required after freezing
  • Thickness, 2.25 to 4 mil
  • RNase, DNase, pryogen and BPH free
  • Comply with regulatory and industrial requirements of EPA, FDA, HACCP and USDA
  • Bags should not be used at temperatures above 180°F (82°C)
Specifications
DescriptionWrite-on Style
AutoclavableNot Autoclavable
Closure TypeClosing Tab
ColorClear
DisposableSingle-use
MaterialPolyethylene
SterilitySterile
Latex FreeYes
No. per Pack500/Pk.
Outer PackagingCardboard
Protective Inner PackagingPlastic
Quantity500/Pk.
Sterilization MethodEthylene Oxide Gas
Temperature Range (English)-320°F to +180°F
Temperature Range (Metric)-196°C to +82°C
Thickness (English)4 mil
Dimensions (L x W)30 x 19cm
Length (English)12 in.
Width (English)7.5 in.
Product TypeSample Bag with Labeling Area
Capacity (English)55 oz.
Capacity (Metric)1.63 L
Length (Metric)30 cm
Width (Metric)19 cm
Thickness (Metric)0.102 mm
Unit SizePack