SealPAK Tubullar Rollstock Pouches
Actual product may vary
Ampac
SealPAK Tubullar Rollstock Pouches
Create custom-size pouches
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (English)Dimensions (L x W)Material
FIS01-812-26A4.50 mil125 ft. x 16 in.Polyester, Polyethylene
FIS01-812-26C2.50 mil250 ft. x 24 in.Polyester, Polyethylene
FIS01-812-26D4.50 mil125 ft. x 9.5 in.Polyester, Polyethylene
FIS01-812-26E2.50 mil250 ft. x 9.5 in.Polyester, Polyethylene
Catalog number FIS01-812-26A
Price (SGD)
3,058.00
Each
Thickness (English):
4.50 mil
Dimensions (L x W):
125 ft. x 16 in.
Material:
Polyester, Polyethylene
Price (SGD)
3,058.00
Each
Create custom-size pouches
  • Ideal for histology and pathology specimens and for safe biological waste disposal in medical, institutional and industrial facilities
  • Polyester with polyethylene lining; does not react with formaldehyde, weak solutions of acids, alkalis or organic solvents
  • Withstand infrared and gamma rays
  • Can be frozen or boiled
  • Can be cut to desired length and sealed at both ends
Specifications
DescriptionHeat-Sealable Roll Stock Pouches
MaterialPolyester, Polyethylene
Thickness (English)4.50 mil
Dimensions (L x W)125 ft. x 16 in.
Length (English)125 ft.
Width (English)16 in.
Length (Metric)38 m
Width (Metric)41 cm
Unit SizeEach