▸
Environmental Samplers
▸
General Purpose Sample Bags
▸
Flexibles SealPAK Heavy-Duty Pouches
Actual product may vary
Ampac
Flexibles SealPAK Heavy-Duty Pouches
Ideal for applications requiring long-term storage, sharper objects, or solutions of formaldehyde, acids, alkalies, greases, oils and organic solvents
Have Questions?
Change view
Catalog Number
Dimensions (L x W)
Capacity (English)
Quantity
FIS01-812-25D
6 x 4 in. (15.2 x 10.2cm)
4 oz.
2400/Cs.
FIS01-812-25H
12 x 8 in. (30.5 x 20.3cm)
32 oz.
960/Cs.
2 Options
Catalog number
FIS01-812-25D
Price (SGD)
226.00
Pack
Add to cart
Dimensions (L x W):
6 x 4 in. (15.2 x 10.2cm)
Capacity (English):
4 oz.
Quantity:
2400/Cs.
Price (SGD)
226.00
Pack
Add to cart
Ideal for applications requiring long-term storage, sharper objects, or solutions of formaldehyde, acids, alkalies, greases, oils and organic solvents
Boilable and freezable pouches can sustain temperatures from -56 to 115°C (-70 to 240°F)
4.5 mils thick
Transparent, airtight and leak-proof
Can be sterilized with gamma, infrared and e-beam
Sealed pouches can also be surface sterilized using VHP
Specifications
Color
Transparent
Material
Polyester
Quantity
2400/Cs.
Thickness (English)
4.50 mil
Dimensions (L x W)
6 x 4 in. (15.2 x 10.2cm)
Length (English)
6 in.
Width (English)
4 in.
Capacity (English)
4 oz.
Length (Metric)
15.2 cm
Width (Metric)
10.2 cm