Round Bottom Sterile Bag for Stomacher™ Lab Blenders
Actual product may vary
Round Bottom Sterile Bag for Stomacher™ Lab Blenders
Round Bottom Sterile Bag for Stomacher™ Lab Blenders
Nasco
Round Bottom Sterile Bag for Stomacher™ Lab Blenders
Round bottom specialty filter bag features a write-on label to make identification quick and easy.
Have Questions?
Catalog NumberCapacity (Metric)
FIS01-8121538 mL
Catalog number FIS01-812
Price (SGD)
853.00
Pack
Capacity (Metric):
1538 mL
Price (SGD)
853.00
Pack
  • Round bottom minimizes sample fragments from getting caught in corners of the bag during blending
  • Made from polyethylene
Specifications
AutoclavableNot Autoclavable
Closure TypeTape and Wire
ColorClear with White Block
DisposableSingle-use
For Use With (Application)Collect, Store, and Transport Samples
For Use With (Equipment)Stomacher, Blender, Homogenizer
IncludesBag
MaterialLDPE
SterilitySterile
Tubing StyleContinuous perimeter seal with tape and wire closure, full-page filter
Fill Volume52 oz.
Latex FreeYes
No. per Pack500/Pk.
Outer PackagingCardboard
Product LineBlender Bag
Protective Inner PackagingPlastic
Sterilization MethodEthylene Oxide Gas
Temperature Range (English)-320°F to +180°F
Temperature Range (Metric)-196°C to +82°C
Thickness (English)4 mil
Dimensions (L x W)33 x 18.5 cm (13 x 7 in.)
Length (English)13 in.
Width (English)7 in.
Product TypeBlender Bag (Round Bottom)
Capacity (English)52 oz.
Capacity (Metric)1538 mL
Length (Metric)33 cm
Width (Metric)18.5 cm
Thickness (Metric)0.102 mm
Unit SizePack