Color-Coded Biohazard Specimen Transport Bags
Actual product may vary
Color-Coded Biohazard Specimen Transport Bags
Color-Coded Biohazard Specimen Transport Bags
Color-Coded Biohazard Specimen Transport Bags
Color-Coded Biohazard Specimen Transport Bags
Color-coded bag lip for easy identification
Have Questions?
Catalog NumberColorMaterial
FIS01-800-10BluePolyethylene
Catalog number FIS01-800-10
Price (SGD)
81.00
Case
Color:
Blue
Material:
Polyethylene
Price (SGD)
81.00
Case
  • Measure 6 x 9 in. (15 x 23cm)
  • Printed with orange biohazard symbol, blue stat and storage instructions
  • Document pouch keeps paperwork separate from specimens
Specifications
AutoclavableNot Autoclavable
Closure TypeZip Seal
ColorBlue
MaterialPolyethylene
SterilityNon-sterile
Dimensions (L x W)9 x 6 in. (23 x 15cm)
Length (English)9 in.
Width (English)6 in.
Length (Metric)23 cm
Width (Metric)15 cm
Unit SizeCase