▸
Facility Safety and Maintenance
▸
Antistatic Storage and Transport
▸
Moisture Barrier Bag, 3.6 mil
Actual product may vary
Moisture Barrier Bag, 3.6 mil
Moisture Barrier Bags, 3.6 mil, protect sensitive electronics from moisture and static damage during storage and transportation.
Have Questions?
Change view
Catalog Number
Dimensions (L x W)
FIS01-800-07
18 x 16 in.
FIS01-800-01
28 x 6 in.
FIS01-800-06
30 x 10 in.
3 Options
Catalog number
FIS01-800-07
Price (SGD)
90.00
Case
Add to cart
Dimensions (L x W):
18 x 16 in.
Price (SGD)
90.00
Case
Add to cart
Protects electronics from moisture and static damage
Opaque and light tight to offer additional protection
Firm lamination and heat sealing offers superior resistance
These bags are ideal for transporting and storing sensitive devices such as circuit boards and electronic components.
Flexible structure and easy to vacuum seal
Specifications
Autoclavable
Not Autoclavable
Closure Type
None
Color
Silver
For Use With (Application)
Packaging of ESD and Moisture Sensitive Electronic Devices
Includes
Bag Only
Quantity
100 Pk.
Thickness (English)
3.6 mil
Dimensions (L x W)
18 x 16 in.
Length (Metric)
457 mm
Width (Metric)
406 mm
Thickness (Metric)
92 μ