Softening Point Apparatus
Softening Point Apparatus
Actual product may vary
Softening Point Apparatus
Softening Point Apparatus
Softening Point Apparatus
Apparatus, Softening Point; Humboldt; With beaker; Shouldered ring-and-ball; Consists of beaker with shelving, two std. balls, two rings, cover; Samples held within horizontal rings; Steel balls placed on top of samples
Have Questions?
Catalog number FIS01-551
Price (SGD)
529.00
EA
Add to cart
Price (SGD)
529.00
EA
Add to cart
Apparatus, Softening Point; Humboldt; With beaker; Shouldered ring-and-ball; Consists of beaker with shelving, two std. balls, two rings, cover; Samples held within horizontal rings; Steel balls placed on top of samples
In accordance with ASTM D 36, E 28, and D 2398 tests
Shouldered ring-and-ball apparatus for softening point determinations according to ASTM D 36, E 28, and D 2398. Apparatus consists of beaker with shelving, balls, rings, cover. Samples are held within horizontal rings. Steel balls are placed on top of samples. As sample, which is submerged in water, is heated, weight of ball forces softened sample downward. Softening point is temperature at which sample falls 1 in. (2.5cm) to bottom shelf. Apparatus comes with two standard balls, two 01-571 shouldered rings, 02-540N beaker, two shelves (1 in. [2.5cm] apart) with support rods, metal cover and 01-572-5 ball retaining rings.
Specifications
Product TypeSoftening Point Apparatus
Certifications/ComplianceMeet ASTM D36, E28, AASHTO T53
For Use With (Application)Brass ring and ball method (double)
Includes800 mL beaker, 2 standard balls, 2 shouldered rings and 2 ball centering guides
Unit SizeEA