Search Fisher Scientific
| Catalog Number | Closure Type | Size | Style |
|---|---|---|---|
| FIS01-369-2B | Buttons | 34 | Knee-length |
| FIS01-369-2A | Buttons | 32 | Knee-length |
| FIS01-369-2C | Buttons | 36 | Knee-length |
| FIS01-369-2E | Buttons | 40 | Knee-length |
41 in. length based on size 40, 100% Sanforized Cotton Heavyweight Twill
420 Series