Tyvek™ 400 Sleeve
Actual product may vary
DuPont
Tyvek™ 400 Sleeve
Nonwoven Tyvek™ fabric helps resist penetration by particles and other contaminants. The inherent barrier stops submicron particles even after abrasion.
Have Questions?
Catalog NumberSeam Type
FIS01-361-21Serged
Catalog number FIS01-361-21
Price (SGD)
375.00
Case
Seam Type:
Serged
Price (SGD)
375.00
Case
  • Elastic at both ends (bicep and wrist) for arm protection
  • 18 in. long
  • Manufactured under specifications that do not contain natural rubber latex
  • One size fits most
  • D13397569
Specifications
Product TypeGeneral Purpose Sleeve
SizeUniversal
MaterialTyvek
ColorWhite
SterilityNon-sterile
DisposableSingle-use
Closure TypeElastic Ends
Seam TypeSerged
For Use With (Application)Lead and Asbestos Abatement/Remedation, General Maintenance/Operations, Spray Painting, General Clean-Up
Length (English)18 in.
Length (Metric)45.7 cm
Packaging100 Cs.
Unit SizeCase