Tyvek™ 400 Lab Coat
Actual product may vary
Tyvek™ 400 Lab Coat
Tyvek™ 400 Lab Coat
DuPont
Tyvek™ 400 Lab Coat
Limited-use lab coats offer easy donning and doffing and provide an ideal balance of protection, durability, and comfort.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizePackaging Type
FIS01-361-10ASmallBulk Packaged
FIS19-141-7625X-LargeBulk Packaged
Catalog number FIS01-361-10A
Price (SGD)
569.00
Case
Size:
Small
Packaging Type:
Bulk Packaged
Price (SGD)
569.00
Case
Limited-use lab coats offer easy donning and doffing and provide an ideal balance of protection, durability, and comfort.
  • Labcoat offers front snap closures for easy donning and doffing
  • Provides enhanced frontal protection
  • Laydown collar
  • Open wrists
  • Two front pockets for holding writing instruments, small tools or parts
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Product TypeGeneral Purpose Lab Coat
SizeSmall
GenderUnisex
MaterialTyvek
ColorWhite
Closure TypeSnaps
Cuff StyleOpen
DisposableLimited-use
SterilityNon-sterile
ResistanceNon-hazardous Liquids
StyleBelow Hip-length
No. of Pockets2
Collar TypeLay Down
ModelTY212SWH
Seam TypeSerged
Size (English) Chest32.25 to 35.75 in.
For Use With (Application)Lead and Asbestos Abatement, Remediation, General Maintenance, Spray Painting, Cleaning
Packaging TypeBulk Packaged
Length (English) Sleeve30.5 in.
Length (Metric) Sleeve77.47 cm
Unit SizeCase