Check Order Status
Reusable Small Animal Feeding Needles: Straight
Actual product may vary
Reusable Small Animal Feeding Needles: Straight
Precision-assembled stainless-steel feeding needles ideal for feeding and dosing
Have Questions?
Catalog Number
Diameter (English) Ball
Gauge
Length (English)
FIS01-290-3A
0.088 in.
20G
1 in.
Catalog number
FIS01-290-3A
Price (SGD)
2,401.00
Each
Add to cart
Diameter (English) Ball:
0.088 in.
Gauge:
20G
Length (English):
1 in.
Price (SGD)
2,401.00
Each
Add to cart
For the intubation of laboratory animals
Straight, curved tubing styles
Many standard sizes available, from small to large
Straight
Autoclavable
Small Animal Needles
For animals 0.7 oz. to 1 lb., 5 oz. (20 to 600g)
Specifications
Autoclavable
Autoclavable
Product Type
Straight Animal Feeding Needle
Material
Stainless Steel
Disposable
Reusable
For Use With (Application)
Small animal, Ideal for feeding and dosing
Gauge
20G
Length (English)
1 in.
Length (Metric)
0.254 cm
Diameter (English) Ball
0.088 in.
Diameter (Metric) Ball
0.023 cm