Two-Color Wash Bottles, Safety-Vented™ and Safety Labeled, Wide-Mouth
Actual product may vary
Bel-Art
Two-Color Wash Bottles, Safety-Vented™ and Safety Labeled, Wide-Mouth
Unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know safety-labeled wash bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Chemical Label
FIS01-189-0104500 mLIsopropanol
FIS11-391-53250 mLAcetone
FIS11-391-55250 mLIsopropanol
FIS11-391-54250 mLEthanol
Catalog number FIS01-189-0104
Price (SGD)
113.00
Pack
Capacity (Metric):
500 mL
Chemical Label:
Isopropanol
Price (SGD)
113.00
Pack
A unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know Safety-Labeled Wash Bottles
  • Pre-printed with chemical name and formula, the National Fire Protection Association (NFPA) section 704 four-color diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
  • Valve eliminates drips, allows dispensing of wash bottle in an upright or inverted position.
  • Wide mouth 53 mm (2 in.) bottle for easy refilling.
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification.
  • Translucent, LDPE solvent-resistant Sodium Hypochlorite is white LDPE for light-protection between 230 and 450.
  • Toluene is red LDPE bottle.
  • Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
  • Labeled with six of the most commonly used solvent names or custom labeling available.
Specifications
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent
Diameter (Metric) Outer Neck53 mm
For Use With (Application)Isopropanol
IncludesYellow Polypropylene Cap
MaterialPolyethylene
Quantity1 Ea.
SterilityNon-sterile
Closure ColorYellow
Certifications/ComplianceOSHA 29 CFR 1910.1200
Closure IncludedYes
DisposableReusable
GraduatedNo
MouthWide
ShapeCylindrical
Product TypeWash Bottle
Chemical LabelIsopropanol
Closure MaterialPolypropylene
Closure TypeSafety Vented Cap