Acid Neutralization Trap for Vacuum Pumps
Actual product may vary
Welch
Acid Neutralization Trap for Vacuum Pumps
Accessory for Welch Vacuum Pump Models 1402N, 1376N
Have Questions?
Catalog NumberSize
FIS01-104-16Up to 2 cfm (57 L/min.)
Catalog number FIS01-104-16
Price (SGD)
1,725.00
Each
Size:
Up to 2 cfm (57 L/min.)
Price (SGD)
1,725.00
Each
For use with Welch Vacuum Pump Models 1402N, 1376N
  • Contains large alkaline element cartridge to neutralize hydrous acids before they enter vacuum pump
  • Transparent body
  • Capacity: Up to 2 cfm and 12 cfm
  • 20.3W x 33cm H
Specifications
For Use WithModel 1402N, 1376N vacuum pump
Overall Height (English)8.97 in.
Overall Height (Metric)22.78 cm
Overall Width (English)7.99 in.
SizeUp to 2 cfm (57 L/min.)
TypeAcid Neutralization Trap
Unit SizeEach