Standard-Duty Vacuum Pumps
Actual product may vary
Welch
Standard-Duty Vacuum Pumps
Compact, economical WOB-L™ pumps for aqueous applications
Have Questions?
Catalog NumberModelFlow Rate
FIS01-051-1A252222 L/min.
Catalog number FIS01-051-1A
Price (SGD)
2,589.00
Each
Model:
2522
Flow Rate:
22 L/min.
Price (SGD)
2,589.00
Each
Compact, economical WOB-L™ pumps for aqueous applications
  • Ideal for filtration, vacuum ovens, aspiration and chamber roughing
  • Protect against accidental ingestion
  • Wetted parts resist corrosion from moisture
Specifications
Horsepower0.12 hp
Shipping Weight (English)17 lb.
Shipping Weight (Metric)7.7 kg
IncludesLiquid trap on inlet, vacuum gauge and adjustment knob, exhaust muffler, suction cup feet and handles for convenient transportation
Certifications/ComplianceCE
Electrical Requirements115 V 60 Hz
Model2522
Weight (English)11.7 lb.
Weight (Metric)5.3 kg
Flow Rate22 L/min.
Frequency60 Hz
Overall Height (English)10 in.
Overall Height (Metric)25.4 cm
Overall Length (English)8.1 in.
Overall Length (Metric)20.6 cm
Overall Width (English)8.8 in.
Maximum Pressure100 psi
Voltage115 V
Unit SizeEach