▸
Pumps
▸
Drum Pumps
▸
Plastic Siphon Drum Pumps
Actual product may vary
Fisherbrand
Plastic Siphon Drum Pumps
Choice of polyethylene or polypropylene models
Have Questions?
Change view
Catalog Number
Flow Rate
Type
FIS01-007-2
5 gal./min.
Buttress Siphon Pump
FIS01-007-13
8 oz./stroke
Drum Pump
2 Options
Catalog number
FIS01-007-2
Price (SGD)
-
Flow Rate:
5 gal./min.
Type:
Buttress Siphon Pump
Choice of polyethylene or polypropylene models
For dispensing of dilute acids, caustics, oils, waxes, disinfectants without leakage problems.
Maximum delivery rate varies with model
Use for 15-30 and 55 gal. (57-114, 208L) drums with a 2 in. (5.1cm) bung
Some units can be used with 5 gal. pail with separate adapter
Heavy-duty model available
Specifications
Applications
Dispensing of dilute acids, caustics, oils, waxes, disinfectants without leakage problems
Flow Rate
5 gal./min.
For Use With
15 to 30 or 55 gal. drum and 5 gal. pail with separate adapter
Type
Buttress Siphon Pump
Temperature (English) Min
40°F
Temperature (English) Max.
140°F
Material
Polyethylene