Plastic Siphon Drum Pumps
Plastic Siphon Drum Pumps
Actual product may vary
Plastic Siphon Drum Pumps
Plastic Siphon Drum Pumps
Fisherbrand
Plastic Siphon Drum Pumps
Choice of polyethylene or polypropylene models
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With
FIS01-007-13Drum Pump15 to 30 or 55 gal. drum and 5 gal. pail with separate adapter
FIS01-007-2Buttress Siphon Pump15 to 30 or 55 gal. drum and 5 gal. pail with separate adapter
Catalog number FIS01-007-13
Price (SGD)
0.00
EA
Add to cart
Type:
Drum Pump
For Use With:
15 to 30 or 55 gal. drum and 5 gal. pail with separate adapter
Price (SGD)
0.00
EA
Add to cart
Choice of polyethylene or polypropylene models
  • For dispensing of dilute acids, caustics, oils, waxes, disinfectants without leakage problems.
  • Maximum delivery rate varies with model
  • Use for 15-30 and 55 gal. (57-114, 208L) drums with a 2 in. (5.1cm) bung
  • Some units can be used with 5 gal. pail with separate adapter
  • Heavy-duty model available
Specifications
ApplicationsDispensing of dilute acids, caustics, oils, waxes, disinfectants without leakage problems
For Use With15 to 30 or 55 gal. drum and 5 gal. pail with separate adapter
Overall Length (English)41 in.
Overall Length (Metric)104.14 cm
Temperature (English) Max.140°F
Temperature (English) Min40°F
MaterialPVC
TypeDrum Pump
Flow Rate8 oz./stroke
Weight (English)2 lb.
Weight (Metric)0.9 kg
Unit SizeEA