Check Order Status
FC400 Flask Cap
FC400 Flask Cap
Savant™ FC400 Flask Cap is suitable for the GCF400 Glass Condensation Flask and improves the functionality of your Savant™ SpeedVac Concentrator.
Have Questions?
Catalog number
FC400
also known as AVL
Price (SGD)
-
Suitable for GCF400 Glass Condensation Flask
For use with SpeedVac Concentrator
Specifications
Color
Black
Description
Flask Cover for GCF400 Flask
Type
Flask Cap
For Use With (Equipment)
GCF400 Flask
Model
FC400