Check Order Status
Bulk tubes
Actual product may vary
Bulk tubes
For use with Bead Mill 4 and Bead Mill 24 homogenizers. Also fits most other commercially available Bead Mills.
Have Questions?
Change view
Catalog Number
Product Type
FBO15545809
Reinforced tubes with screw caps and O-rings
FBO15535809
Non-reinforced tubes with screw caps and O-rings
2 Options
Catalog number
FBO15545809
Price (SGD)
1,197.00
EA
Add to cart
Product Type:
Reinforced tubes with screw caps and O-rings
Price (SGD)
1,197.00
EA
Add to cart
RNase/DNase free
Processing range: 300μL to 1.8mL
Beads not included
Specifications
Product Type
Reinforced tubes with screw caps and O-rings
Capacity (Metric)
2 mL
Color
White/Opaque
Dnase Free
Yes
For Use With (Equipment)
Bead Mill 4 and 24 Homogenizers
Packaging Quantity
500/Pk.
RNase Free
Yes