1x1 image pixel for data collection
150 Homogenizer Accessory, Motor Stand Assembly
150 Homogenizer Accessory, Motor Stand Assembly
Actual product may vary
150 Homogenizer Accessory, Motor Stand Assembly
Fisher Scientific™ 150 Homogenizer Accessory, Motor Stand Assembly designed to retain the Fisher Scientific 150 Homogenizer for post mounted operation.
Have Questions?
Catalog NumberFor Use With (Equipment)
FBO15525819Fisher Scientific 150 Homogenizer
Catalog number FBO15525819
Price (SGD)
786.00
EA
Add to cart
For Use With (Equipment):
Fisher Scientific 150 Homogenizer
Price (SGD)
786.00
EA
Add to cart
  • Includes a baseplate and post
  • Base dimensions: 12.7 x 20.3cm (5 x 8 in.) stainless steel base
  • Post dimensions: 48.2cm (19 in.) tall by 9.52mm (0.375 in.) diameter stainless steel support post
Specifications
Product TypeMotor Stand Assembly
MaterialStainless Steel
For Use With (Equipment)Fisher Scientific 150 Homogenizer
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection