Product Image
Eyela
Roll Form European Standard Pipe Marker with Die-cut Arrowheads - Combustible Liquids
Have Questions?
Catalog NumberDisplay Text
EYE229320Hydraulic Oil
Catalog number EYE229320
Price (SGD)
485.00
Each
Display Text:
Hydraulic Oil
Price (SGD)
485.00
Each
Specifications
Certifications/ComplianceEuropean Directives 67/548 and 88/379
DescriptionRoll form Pipe Markers with liner, without pictograms - Flammable/Non Flammable Liquids/Oils
Adhesive TypePermanent Acrylic
ColorWhite, Brown
FinishGloss
MaterialPolyester
Product TypeRoll Form Pipe Marker
Display TextHydraulic Oil
For Use With (Application)Used for pipe markers and safety signs
LanguagesEnglish
Quantity220/Pk.
Temperature Range (Metric)-40°C to +110°C
Thickness (Metric)0.099 mm
Dimensions (L x W)150 x 100 mm
Width (Metric)100 mm
Height (Metric)150.00 mm
Height (Metric) Character13 mm
Unit SizeEach