Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Foot Protection
▸
General Purpose Work Boots and Shoes
▸
Brown Nubuck Safety Boot Dual Density Sole & Midsole
Eyela
Brown Nubuck Safety Boot Dual Density Sole & Midsole
Brown safety shoe constructed of water resistant leather with a dual density PU sole and steel midsole.
Have Questions?
Catalog Number
Size
EYE120110
10
Catalog number
EYE120110
Price (SGD)
90.00
Pack
Add to cart
Size:
10
Price (SGD)
90.00
Pack
Add to cart
Quality leather uppers
Water resistant leather
Dual density PU sole
Steel midsole
Anti-slip and Anti-static
ENISO 20345-1 S3
Specifications
Color
Brown
Material
Nubuck
Size
10
Sole Features
Dual Density