7000 Series Ultra Precision Wire Cutters with Medium Heads
7000 Series Ultra Precision Wire Cutters with Medium Heads
Actual product may vary
7000 Series Ultra Precision Wire Cutters with Medium Heads
For cutting leads and soft wire
Have Questions?
Catalog NumberCapacity (English)Cut TypeHandle TypeHead Style
ECT7248E0.005 to 0.036 in.Optimum FlushErgonomic shortMedium Tapered Relieved
Catalog number ECT7248E
Price (SGD)
201.00
EA
Add to cart
Capacity (English):
0.005 to 0.036 in.
Cut Type:
Optimum Flush
Handle Type:
Ergonomic short
Head Style:
Medium Tapered Relieved
Price (SGD)
201.00
EA
Add to cart
  • Induction hardened carbon tool steel
  • Edges hardened to RHC 63-65
  • Dual leaf springs
  • ESD safe cushion grips
Specifications
Handle TypeErgonomic short
GradeUltra Precision
Head StyleMedium Tapered Relieved
Length (English)5 in.
Length (Metric)12.7 cm
Material1% Carbon Tool Steel
Capacity (English)0.005 to 0.036 in.
Cut TypeOptimum Flush
Grip TypeESD-safe soft foam cushion
IncludesMedium tapered/relieved, set-screw, induction hardened cutting blades to RHC 63-65, Stainless Steel dual leaf springs
Length (English) Head0.44 in.
Length (English) Tip0.32 in.
Length (Metric) Head1.11 cm
Length (Metric) Tip0.81 cm
Width (English) Head0.05 in.
Width (Metric) Head0.12 cm
Unit SizeEA