Scratch Brush Refills
Scratch Brush Refills
Actual product may vary
Scratch Brush Refills
Bristles for presolder and contact cleaning
Have Questions?
Catalog NumberDiameter (English) BristleBristle MaterialSize
ECT267A0.13 in.Fiberglass0.125 in. (3.18 mm)
Catalog number ECT267A
Price (SGD)
4.00
EA
Add to cart
Diameter (English) Bristle:
0.13 in.
Bristle Material:
Fiberglass
Size:
0.125 in. (3.18 mm)
Price (SGD)
4.00
EA
Add to cart
  • Fine precision
  • Fine deburring
  • Polishing
  • Rust removal
  • Cleaning and erasing in chemical etching processes
Specifications
Product TypeBrush Refill
Bristle MaterialFiberglass
Diameter (English) Bristle0.13 in.
Diameter (Metric) Bristle0.33 cm
For Use With (Application)Fine deburring, polishing, rust removal, cleaning and erasing in chemical etching processes
For Use With (Equipment)Scratch brush
Length (English)0.125 in.
Length (Metric)0.317 cm
PrecisionFine
Size0.125 in. (3.18 mm)
Unit SizeEA