Thermo Scientific
Eutech™ KCl Calibration Solution
For conductivity calibration and standards, choose from our available Eutech™'s Conductivity Standards, Calibrations, and Solutions
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConductivity
ECCON84BT480 mL84 μS/cm
FKS15488972480 mL12.88 mS/cm
ECCON1118BT480 mL111.8 mS/cm
Catalog number ECCON84BT
HSN 90318000
Price (SGD)
-
Quantity:
480 mL
Conductivity:
84 μS/cm
Eutech™ KCl Calibration Solutions are used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.
Features:
  • Packed in 60mL or 480 ml durable plastic bottles
  • Economical and convenient to use
Specifications
Description84 μS/cm KCl calibration solution
Conductivity84 μS/cm
pH7
Boiling Point100°C
For Use With (Equipment)Conductivity Meters and Electrodes
SolubilitySoluble
Solution TypeCalibration Solution
TypeConductivity Standard
Industry TypeEconomical and Convenient for Laboratory Use
Quantity480 mL
Unit SizeEach