Nalgene™ Centrifuge Tube Sealing Cap Assemblies
Nalgene™ Centrifuge Tube Sealing Cap Assemblies
Actual product may vary
Nalgene™ Centrifuge Tube Sealing Cap Assemblies
Nalgene™ Centrifuge Tube Sealing Cap Assemblies
Nalgene™ Centrifuge Tube Sealing Cap Assemblies
Nalgene™ Centrifuge Tube Sealing Cap Assemblies
Process tubes at their highest rated speed level (10,000 x g or greater) without leakage, even when full with these sealing cap assemblies.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
DS3131-0024Closure size 24
FKS10383341Closure size 24
Catalog number DS3131-0024
Price (SGD)
40.00
CS
Add to cart
For Use With (Equipment):
Closure size 24
Price (SGD)
40.00
CS
Add to cart
Process Thermo Scientific™ Nalgene™ FEP Oakridge 30mL and 50mL centrifuge tubes at their highest rated speed level (10,000 x g and over) without leakage, even when full. Thermo Scientific™ Nalgene™ Sealing Cap Assemblies include a Viton™ fluoroelastomer o-ring. Also recommended when spinning hazardous materials compatible with centrifuge tube material.
Features:
  • Top fits securely for use at maximum speeds without leakage
  • Autoclavable/Biohazard/Leakproof† when used with Nalgene FEP Oakridge tubes
Note: Not recommended for use with centrifuge tubes made from materials other than FEP.
Includes:
  • Polypropylene screw closure
  • Tefzel™ ETFE plug
  • Viton™ fluoroelastomer o-ring
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Closure Size24 mm
Product TypeCentrifuge Tube Cap
AutoclavableAutoclavable
DescriptionSealing cap assembly
MaterialPolypropylene, ETFE, Fluoroelastomer
No. per Pack2
No. per Case2
Product LineNalgene™
For Use With (Equipment)Closure size 24
Unit SizeCS
Frequently asked questions (FAQs)
Are there replacement O rings for Nalgene Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap?
Replacement sealing-cap assemblies are available for selected sizes. We recommend Nalgene Centrifuge Tube Sealing Cap Assemblies (Cat. No. DS3131-0020) for 30 mL tubes and Nalgene Centrifuge Tube Sealing Cap Assemblies (Cat. No. DS3131-0024) for 50 mL tubes.
The actual brim capacity of the Nalgene Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap is 42 mL, but it is designed to fit a standard rotor.