Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes
Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes
Actual product may vary
Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes
Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes
Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes
Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes
Use these friction-fit polypropylene closures featuring a convenient pull tab for easy removal from centrifuge tubes.
Have Questions?
Catalog NumberFor Use With (Equipment)
DS3111-0032Tube with 32 mm O.D.
Catalog number DS3111-0032
Price (SGD)
73.00
CS
Add to cart
For Use With (Equipment):
Tube with 32 mm O.D.
Price (SGD)
73.00
CS
Add to cart
Easily remove Thermo Scientific™ Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes from the centrifuge with a convenient pull tab. These autoclavable closures are designed to fit Nalgene™ round and conical centrifuge tubes. They may not be liquid-tight during high-speed centrifugation.
Use with Nalgene conical and round bottom open top centrifuge tubes.
DS3111-0030 fits non-lipped 50mL centrifuge tubes only.
Ordering Information:
  • Nalgene 15mL conical centrifuge tubes use the 17mm closure
  • Nalgene 50mL conical centrifuge tubes use the 29mm closure
  • Nalgene 12ml round bottom centrifuge tubes use the 16mm closure
  • Nalgene 15ml round bottom centrifuge tubes use the 16mm closure
  • Nalgene 16ml round bottom centrifuge tubes use the 18mm closure
  • Nalgene 38ml round bottom centrifuge tubes use the 25mm closure
  • Nalgene 50ml round bottom centrifuge tubes use the 29mm closure
  • Nalgene 100ml round bottom centrifuge tubes use the 32mm closure
Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) centrifuge tubes are designed to last many spins under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These centrifuge tubes are recyclable in many communities. At end of product life, please wash and recycle your Nalgene PP and PPCO centrifuge tubes if services are available in your community.
Specifications
Closure Size32 mm
Product TypeCentrifuge Tube Cap
AutoclavableAutoclavable
DescriptionDesigned to be compatible with Nalgene round and conical centrifuge tubes
MaterialPolypropylene
No. per Pack20
No. per Case20
Product LineNalgene™
For Use With (Equipment)Tube with 32 mm O.D.
Unit SizeCS