Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Actual product may vary
Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Freeze samples effectively with impact-resistant these narrow-mouth, polycarbonate bottles with closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
DS2205-00104 L38-430 mm
2205-0016
also known as 2205-0016JP
0.5 L38-430 mm
2205-0032
also known as 2205-0032C
1 L38-430 mm
FIS02-960-52C2 L38-430 mm
Catalog number DS2205-0010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
4 L
Closure Size:
38-430 mm
Freeze samples effectively with impact-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure, which are window clear. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features:
  • 16 oz. and 0.25 gal. sizes are good for smaller volume applications where contents need to be seen
  • 0.5 and 0.66 gal. sizes can be used as culture vessels for batch, suspension or nonattached roller culturing
  • 1 and 2 gal. sizes have built-in shoulder loops for attaching an identification tag
  • Excellent impact resistance makes these bottles good for freezer use to -135°C (-211°F)
  • Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable/Transparent/Leakproof† when used with Nalgene closures
Includes:
Leakproof† polypropylene screw caps
Note:
Before autoclaving, completely disengage threads or remove cap. Autoclave empty. Repeated autoclaving will shorten the bottle life.
.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity1 gal., 4L
Capacity (Metric)4 L
Capacity (English)1 gal.
ColorClear
DescriptionNarrow-Mouth Bottle
Diameter (Metric) Inner Neck25 mm
MaterialPolycarbonate
No. per Case1
No. per Pack1
SterilityNon-sterile
Closure ColorNatural
Neck Finish38mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1580 for food and beverage use, USP Class VI.
Closure IncludedYes
AutoclavableAutoclavable
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)333 mm
Product TypeNarrow Mouth Bottle
Width (English)6 in.
Height (English)13.1 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38-430 mm
Diameter (Metric)121 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCS