Barnstead™ ROpure LP™ Prefilters
Product Image
Barnstead™ ROpure LP™ Prefilters
Compatible with Economy Filter Holder
Have Questions?
Catalog NumberPore Size
D27295 μm
Catalog number D2729
Price (SGD)
120.00
EA
Add to cart
Pore Size:
5 μm
Price (SGD)
120.00
EA
Add to cart
Compatible with Economy Filter Holder
  • Length: 10 in. (25.4cm)
  • Can also be used with Economy Filter Holders
Specifications
Flow Rate2
Pore Size5 μm
Feedwater Temperature (English)40°F to 120°F
Feedwater Temperature (Metric)4°C to 49°C
For Use With (Equipment)Economy and Pcs Holders, Prefilter With Economy Filter Holder
TypePrefilter with Economy Filter Holder
Length (English)10 in.
Length (Metric)25.4 cm
Diameter (English)2.5 in.
Diameter (Metric)6.5 cm
Unit SizeEA