MaxiThermal Software
Actual product may vary
MaxiThermal Software
MaxiThermal Software for USB Data Loggers.
Have Questions?
Catalog NumberSoftware Type
CTL15-081-150MaxiThermal
Catalog number CTL15-081-150
Price (SGD)
794.00
Each
Software Type:
MaxiThermal
Price (SGD)
794.00
Each
MaxiThermal Software for USB Data Loggers.
  • Graphic representations are a simple X – Y plot of time versus temperature with unlimited zoom, view temperature data points on the graph, view total elapsed time, or the exact time when it was recorded
  • Graph and all data points (date, time and temperature) can be printed, as well as the ability to add a user note to the file for including information pertinent to that particular plot
  • Ability to view numeric data in a screen window and Data can be Exported as a CSV file for use in other software
  • Summary Information report showing the specific logger information, the Start and Stop Time, Duration of Recording, Measurement Interval, Highest, Lowest, Median and Average Temperatures as well as the High and Low Alarm information
  • Data from the logger is easily downloaded to your PC via a standard USB 1.1 interface
  • Compatible with Windows 98, NT, 2000, ME, XP, Vista, and Windows 7 and 8
Specifications
TypeDatalogger Software
Display TypeNo Display
Software TypeMaxiThermal
IncludesMaxiThermal Software CD
For Use With (Application)Viewing and transferring recorded data from loggers to computer
For Use With (Equipment)Fisherbrand Traceable Datalogging Logger-Trac Thermometers
Unit SizeEach