Traceable™ Logger-Trac™ Temperature Datalogger
Traceable™ Logger-Trac™ Temperature Datalogger
Actual product may vary
Traceable™ Logger-Trac™ Temperature Datalogger
For monitoring material during storage, handling and transportation
Have Questions?
Catalog number CTL15-081-144
Price (SGD)
301.00
EA
Add to cart
Price (SGD)
301.00
EA
Add to cart
Reusable device with active LCD allows cumulative duration of temperature excursion from user-selected MIN/MAX values to be viewed immediately and downloaded if needed. The Fisher Scientific™ Traceable™ Logger-Trac™ Temperature Datalogger is used for monitoring material during storage, handling and transportation.
  • Displays cumulative Alarm Duration from user selected MIN/MAX values
  • Integrates with MaxiThermal Software for complete data analysis
  • Reusable/Re-programmable
  • Size: 2.56 x 1.34 x 0.35 inches
  • Weight: 18.5 grams (0.65 ounces)
  • Storage Capacity: 7,680 readings
  • IP66 Waterproof Rating
  • Individual Logger-Trac Thermometers are pre-programmed with alarm values of 2°C (low) and 8°C (high) and a recording interval to allow for 30 days of recording (readings are logged every 5 minutes, 33 seconds)
Active LCD shows the following important data:
  • Current, MIN/MAX temperatures
  • Recording status
  • Run Time
  • Alarm (Excursion) Cumulative Time
  • Quarantine status
  • Alarm Tally
MaxiThermal™ Software is needed to activate these dataloggers
Specifications
TypeTemperature Datalogger
Display TypeLCD
Data Memory7680 Readings
CalibrationCalibrated
Alarm TypesVisual
Software TypeMaxiThermal
Battery TypeCR2016 3 V Lithium Coin Cell
Battery Lifeone year of constant operating use, 3 years shelf life
IncludesBattery and individually-numbered Traceable Certificate
Certifications/ComplianceCE, Traceable Certificate
IP RatingIP66
MaterialABS Plastic
Sensor TypesWired Chip Glass Thermistor
For Use With (Application)For monitoring material during transportation
Accuracy (Temperature)±0.2°C from 0 to 10°C; ±1°C otherwise
Time Accuracy2 min./month
Temperature Resolution0.1°C
Temperature Range (Metric)-35°C to +50°C
Length (English)2.56 in.
Length (Metric)6.5 cm
Weight (English)0.65 oz.
Weight (Metric)18.5 g
Unit SizeEA