Traceable™ Platinum Ultra-Accurate Digital Thermometer
Actual product may vary
Traceable™ Platinum Ultra-Accurate Digital Thermometer
High-precision unit is designed for years of reliable service—even in the severest environments.
Have Questions?
Catalog NumberTemperature Range (Metric)Model
CTL15-081-102-200°C to +500°CHandle Probe
Catalog number CTL15-081-102
Price (SGD)
1,419.00
Each
Temperature Range (Metric):
-200°C to +500°C
Model:
Handle Probe
Price (SGD)
1,419.00
Each
High-precision unit is designed for years of reliable service—even in the severest environments. Fisher Scientific™ Traceable™ Platinum Ultra-Accurate Digital Thermometers have platinum sensor which ensures linear response and high accuracy across the entire range. Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens and electronics manufacturing.
  • Platinum sensor insures linear response and high accuracy across the entire range
  • Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens, electronics manufacturing
  • Membrane-covered keys are splash-proof and easy to operate
  • HOLD allows user to freeze display for reading later
  • Displays data in °F or °C
  • Computer output supplied with unit
  • Handle Probe: (range –200 to 500°C): Fast-response, waterproof, 4-wire platinum/stainless-steel 316 probe, 0.16-inch diameter x 7-inch stem length, overall length 11.5 inches; probe tip penetration of 0.3-inch, 6-foot cable
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Depth (English)0.5 in.
Depth (Metric)1.27 cm
Diameter (English) Probe0.16 in.
Diameter (Metric) Probe0.4 cm
Dimensions (L x W x H)11.5 x 3.5 x 5.5 in. (29.21 x 8.89 x 13.97 cm)
For Use With (Application)Measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater
Height (English)5.5 in.
Height (Metric)13.97 cm
IncludesStainless Steel probe with handle, cable, computer output capabilities (software package must be purchased separately), Plastic carrying case, traceable certificate and 9V battery
Length (English) Cable6 ft.
Length (English) Probe7 in.
Length (Metric)292 mm
Length (Metric) Cable182.88 cm
Length (Metric) Probe17.7 cm
MaterialPlatinum, Stainless Steel
ModelHandle Probe
Resolution0.001°C
Weight (English)8 oz.
Weight (Metric)226.7 g
Width (English)3.5 in.
Product TypeDigital Thermometer
Length (English)11.5 in.
Max. Temperature (English)932°F
Max. Temperature (Metric)500°C
Min. Temperature (English)-328°F
Min. Temperature (Metric)-200°C
Temperature Range (Metric)-200°C to +500°C
CalibratedYes
Accuracy±0.05°C
DisplayDigital LCD
Width (Metric)8.89 cm
Unit SizeEach