1x1 image pixel for data collection
Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Actual product may vary
Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Traceable™ Digital Thermometer used to measure temperature of gases, food, and semisolids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)Temperature Range (English)AccuracyModel
CTL15-078K-50°C to +150°C-58°F to +302°F±1° (-20° and 100°C)Oven and Incubator Thermometer
CTL15-077-58-50°C to +150°C-58°F to +302°F±1°CKey-Chain Thermometer
CTL14-648-44-50°C to +300°C-58°F to +572°F±1°CShockproof/Water-resistant Thermometer
CTL14-648-43-50°C to +280°C-58°F to +536°F±1°CFood/Piercing/Splash-Proof/Drop-Proof Thermometer
CTL06-664-27-50°C to +300°C-58°F to +572°F±0.4°CUltra Shockproof/Water-resistant Thermometer
CTL06-664-35-50°C to +300°C-58°F to +572°F±0.4°CUltra Food/Waterproof/Piercing/Drop-proof Thermometer
Catalog number CTL15-078K
Price (SGD)
107.00
EA
Add to cart
Temperature Range (Metric):
-50°C to +150°C
Temperature Range (English):
-58°F to +302°F
Accuracy:
±1° (-20° and 100°C)
Model:
Oven and Incubator Thermometer
Price (SGD)
107.00
EA
Add to cart
Traceable™ Digital Thermometer used to measure temperature of gases, food, and semisolids.
  • Short-stem piercing models are ideal for carrying in a pocket
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
  • Traceable to standards provided by NIST
  • Waterproof thermometers (see Models 14-648-43, -44) are ideal for wet labs, wash-down areas and field use
  • 0.375 in. LCD screen thermometers available (see Cat. No.14-648-47); also available with protective guards (see Cat. Nos. 06-664-32 and 06-664-30)
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Depth (English)Not Applicable
Depth (Metric)Not Applicable
Diameter (English) Probe0.12 in.
For Use With (Application)Measuring temperature in soil, cuvettes, test tubes, food products, incubators and petroleum hydrocarbons
IncludesStainless steel probe and Protective case
Length (English) Probe8 in.
Length (Metric)280 mm
Length (Metric) Probe20 cm
MaterialStainless Steel
ModelOven and Incubator Thermometer
Resolution0.1° (-20°C to +200°C)
Product TypeDigital Thermometer
Length (English)11 in.
Max. Temperature (English)302°F
Max. Temperature (Metric)150°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +302°F
Temperature Range (Metric)-50°C to +150°C
CalibratedYes
Accuracy±1° (-20° and 100°C)
DisplayDigital LCD
Width (Metric)3 cm
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link