Traceable™ ISO 17034 (formerly ISO Guide 34) Color-Coded CRM pH Buffers (Certified Reference Material), Fisherbrand™
Traceable™ ISO 17034 (formerly ISO Guide 34) Color-Coded CRM pH Buffers (Certified Reference Material), Fisherbrand™
Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers.Compatible with all makes of pH instruments and electrodes.
Have Questions?
Change viewbuttonViewtableView
pH:
7.000
4.005
10.012
Color:
View all
Yellow
Catalog number CTL15-078-281
Price (SGD)
68.00
Each
pH:
7.000
Color:
Yellow
Request bulk or custom format
Price (SGD)
68.00
Each
Specifications
Chemical Name or MaterialBuffer Standards pH 7.000(CRM)
ColorYellow
For Use With (Application)Calibration
For Use With (Equipment)pH Instruments and Electrodes
pH7.000
View more
Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.
  • Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
  • 100% compatible with all makes of pH instruments and electrodes
  • Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
  • Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
  • Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
  • Each 16 ounce bottle or six-pack single-use 100ml containers is supplied with individual temperature chart, traceability information and Traceable™ Certificate