Traceable™ Wide-Range Thermometer
Actual product may vary
Traceable™ Wide-Range Thermometer
Traceable™ Wide-Range Thermometer is certified Traceable for ISO 9000 and other requirements.
Have Questions?
Catalog NumberLength (English)
CTL15-078-22.75 in.
Catalog number CTL15-078-2
Price (SGD)
1,049.00
Each
Length (English):
2.75 in.
Price (SGD)
1,049.00
Each
Traceable™ Wide-Range Thermometer is certified Traceable for ISO 9000 and other requirements.
  • Resolution: 0.1°C or °F from -200° to 640°, otherwise 1°; meter accuracy ±1°C
  • Large, 0.5 in. (1.3 cm) high LCD is easy to read
  • Minimum and maximum readings may be recalled instantly
  • Unit features backlit display and timer
  • Uses J, K, T probes; computer output
  • Supplied with stainless-steel Type-K probe with handle
  • Size: 2.75 x 7 x 1.25 in. (6.98 x 17.78 x 3.17 cm)
  • Weight: 7 oz. (198.44 g)
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
For Use With (Application)Measuring Temperature in Liquids, Air/Gas and Semi-solids
Height (English)1.25 in.
Height (Metric)3.17 cm
Length (Metric)70 mm
MaterialPlastic
Probe TypeType K Thermocouple
Resolution0.1°C
Weight (English)6.5 oz.
Weight (Metric)184 g
Length (English)2.75 in.
Max. Temperature (English)2200°F
Max. Temperature (Metric)1200°C
Min. Temperature (English)-350°F
Min. Temperature (Metric)-212°C
Accuracy±1°C
DisplayDigital LCD
Unit SizeEach