Traceable™ Anemometer/Thermometers
Actual product may vary
Traceable™ Anemometer/Thermometers
Traceable™ Anemometer/Thermometers recall minimum/maximum readings at the press of a button
Have Questions?
Catalog NumberCase Material
CTL15-078-195ABS Plastic
Catalog number CTL15-078-195
Price (SGD)
519.00
Each
Case Material:
ABS Plastic
Price (SGD)
519.00
Each
Traceable™ Anemometer/Thermometers recall minimum/maximum readings at the press of a button
  • Simple operation
  • Tough ABS plastic housing; durable in severe lab conditions
  • O-ring sealing
  • Membrane keypad keeps out dust and dirt
  • HOLD button freezes the reading on the display
  • High contrast 0.375 in.-high LCD digits
  • Dimensions: 1.50 x 8.25 x 1.50 in. (38.1 x 209.5 x 38.1 mm)
  • Weight: 6 oz. (170.1 g)
Specifications
Accuracy±0.8°C
Case MaterialABS Plastic
Certifications/ComplianceISO 17025, A2LA, NIST
For Use With (Application)Measuring Airflow, Temperature, Humidity and Dew Point
Length (English)8.25 in.
Length (Metric)21 cm
Temperature (English)32°F to 122°F
Temperature (Metric)0°C to 50°C
Weight (English)6 oz.
Weight (Metric)170 g
Unit SizeEach