Traceable™ Big-Digit See-Thru™ Thermometer
Traceable™ Big-Digit See-Thru™ Thermometer
Actual product may vary
Traceable™ Big-Digit See-Thru™ Thermometer
Traceable™ Big-Digit See-Thru™ Thermometer
Traceable™ Big-Digit See-Thru™ Thermometer
Use the Traceable™ Big-Digit See-Thru™ Thermometer outdoors, in storerooms, fume hoods, labs, cleanrooms, refrigerators, and plant areas.
Have Questions?
Catalog NumberTemperature Range (Metric)Temperature Range (English)Scale
CTL15-077-960-25°C to +70°Cn.a.Celsius
Catalog number CTL15-077-960
Price (SGD)
101.00
Each
Add to cart
Temperature Range (Metric):
-25°C to +70°C
Temperature Range (English):
n.a.
Scale:
Celsius
Price (SGD)
101.00
Each
Add to cart
Use the Traceable™ Big-Digit See-Thru™ Thermometer outdoors, in storerooms, fume hoods, labs, cleanrooms, refrigerators, and plant areas.
  • Window mount, wall mount
  • Automatically clears and updates the min./max. readings daily
  • Accuracy: ±1°C
  • Resolution: 0.1°
Specifications
Certifications/ComplianceDNV, ISO 17025, A2LA
Dimensions (L x W x H)4.25 x 3.5 x 0.75 in. (10.8 x 8.9 x 1.9 cm)
For Use With (Application)Outdoors, in storerooms, at fume hoods, labs, clean rooms, refrigerators and plant areas
Height (English)0.75 in.
Height (Metric)1.9 cm
IncludesMounting tape, Velcro™, AAA battery
Length (Metric)108 mm
MaterialPlastic
Resolution0.1°C
Weight (English)8 oz.
Weight (Metric)200 g
Width (English)3.5 in.
Product TypeDigital Thermometer
Length (English)4.25 in.
Max. Temperature (English)158°F
Max. Temperature (Metric)70°C
Min. Temperature (English)-13°F
Min. Temperature (Metric)-25°C
Temperature Range (English)n.a.
Temperature Range (Metric)-25°C to +70°C
CalibratedYes
Battery TypeAAA
Accuracy±1°C
DisplayDigital LCD
Width (Metric)8.9 cm
ScaleCelsius
Unit SizeEach