Specifications
Bottle FillGlycol
Bottle Size5 mL
Certifications/ComplianceISO 17025, A2LA, NIST
Diameter (English) Probe0.3 in.
Dimensions (L x W x H)2.75 x 4.25 x 0.75 in. (7 x 10.7 x 1.9 cm)
For Use With (Application)Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators
Height (English)0.75 in.
Height (Metric)19 mm
IncludesTraceable certificate, flip open stand, wall mounting, Velcro™ and magnetic strips
Length (English) Cable10 in.
Length (English) Probe1.25 in.
Length (Metric)70 mm
Length (Metric) Cable25.4 cm
Length (Metric) Probe3.17 cm
MaterialABS Plastic
ModelRefrigerator/Freezer Thermometer with 5mL bottle
Resolution1°C
Weight (English)4 oz.
Weight (Metric)110 g
Width (English)4.25 in.
Product TypeFridge/Freezer Digital Thermometer
Length (English)2.75 in.
Max. Temperature (English)158°F
Max. Temperature (Metric)70°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +158°F
Temperature Range (Metric)-50°C to +70°C
CalibratedYes
For Use With (Equipment)Refrigerators/Freezers
Battery TypeAA Alkaline (1)
Accuracy±1°C
DisplayDigital LCD
Width (Metric)10.7 cm
Unit SizeEach