Traceable™ Digital Barometer
Actual product may vary
Traceable™ Digital Barometer
Perfect unit for plants and labs to monitor barometric pressure, temperature, relative humidity, barometric pressure trend and time-of-day. Fisherbrand Traceable Digital Barometer is ideal for use in environments where there is a need to monitor conditions of environmental air parameters.
Have Questions?
Catalog NumberAccuracy (Barometer)
CTL14-650-118±4 mbar
Catalog number CTL14-650-118
Price (SGD)
573.00
Each
Accuracy (Barometer):
±4 mbar
Price (SGD)
573.00
Each
Perfect unit for plants and labs to monitor barometric pressure, temperature, relative humidity, barometric pressure trend and time-of-day. Fisherbrand Traceable Digital Barometer is ideal for use in environments where there is a need to monitor conditions of environmental air parameters.
  • A four-line LCD continuously and simultaneously shows barometric pressure, temperature, relative humidity, barometric pressure trend and time
  • Altitude adjustment: -100 to 2500 meters
  • MIN/MAX readings for barometric pressure, temperature and relative humidity are stored every hour for the last 24 hours and are available for recall at the touch of a button
  • Barometric pressure trend shows bar-chart format of the current and past 1/3/6/12 and 24 hour readings
  • Computer output allows unit to be connected to a computer using the accessory Computer Data Acquisition System Software (sold separately, part number 14-650-120) to permanently capture the readings
Specifications
  • Barometric Pressure Range: 500 to 1030mbar
  • Barometric Pressure Resolution: 1mbar
  • Barometric Pressure Accuracy: ±4mbar
  • Temperature Range: 0° to 55°C (32° to 131°F)
  • Temperature Resolution: 0.1°
  • Temperature Accuracy: ±0.4°C
  • Relative Humidity Range: 5 to 95%
  • Relative Humidity Resolution: 1% RH
  • Relative Humidity Accuracy: ±3% RH between 5 to 75%, otherwise ±5% RH
  • Dimensions: 10 x 7.6 x 2.5cm (4 x 3 x 1 in.)
  • Weight: 198g (7 oz.)
Specifications
Accuracy (Barometer)±4 mbar
Humidity Range0 to 95%
Resolution (Humidity)0.01
Resolution (Temperature)0.1°C
Temperature Range (Metric)0°C to 55°C
Range500 to 1030mbar (14.77 to 30.41 in Hg)
Resolution1mbar
Temperature Range (English)32°F to 131°F
Accuracy (Temperature)±0.4°C
Accuracy (Relative Humidity)±3% RH between 5 to 75%, otherwise ±5% RH
Altitude Compensation (Metric)100 to 2500 m
IncludesTraceable Certificate, wall mount, flip open stand and batteries
Certifications/ComplianceIndividually-numbered NIST Traceable™ Certificate assures accuracy from an ISO/IEC 17025:2005 calibration laboratory accredited by A2LA.
TypeDigital Barometer
Weight (Metric)198 g
Weight (English)7 oz.
Unit SizeEach