Traceable™ Digital-Bottle™ Refrigerator Thermometers
Actual product may vary
Traceable™ Digital-Bottle™ Refrigerator Thermometers
Innovative Digital-Bottle Refrigerator/Freezer Thermometers with MINIMUM/MAXIMUM readings
Have Questions?
Catalog NumberBottle FillModel
CTL06-664-255Nontoxic GlycolStandard Model
Catalog number CTL06-664-255
Price (SGD)
184.00
Each
Bottle Fill:
Nontoxic Glycol
Model:
Standard Model
Price (SGD)
184.00
Each
Fisher Scientific™ Traceable™ Digital-Bottle™ Refrigerator Thermometers display minimum and maximum readings over any time period, and entire unit is waterproof
  • Temperature range is -22° to +122°F (-30° to +50°C)
  • Resolution is 0.1°
  • Accuracy is ±1.0°C/Ultra model accuracy is ±0.4°C at tested points
  • Supplied with NIST certificate
  • Stainless-steel probe protects high-accuracy thermistor sensor
  • Liquid-filled GRAS bottle mimics liquid sample temperatures
  • GRAS - liquid generally recognized as safe by U. S. Food and Drug Administration
  • Available with liquid fill or glass bead fill
  • Chemical-resistant ABS plastic holder stands on a shelf or side of holder affixes to a refrigerator wall or any wall with supplied Velcro - is 2.25 x 2.25 x 1.25 in.
Specifications
Bottle FillNontoxic Glycol
Certifications/ComplianceISO 17025, A2LA, NIST
IncludesStainless Steel probe, ABS Plastic holder, double back tape and Velcro™ and replaceable Silver Oxide battery
Length (Metric)117 mm
MaterialABS Plastic
ModelStandard Model
Resolution0.1°C
Weight (English)3.66 oz.
Weight (Metric)102.05 g
Product TypeDigital Thermometer
Length (English)4.63 in.
Max. Temperature (English)122°F
Max. Temperature (Metric)50°C
Min. Temperature (English)-4°F
Min. Temperature (Metric)-20°C
Temperature Range (Metric)-20°C to +50°C
CalibratedYes
For Use With (Equipment)Freezer, refrigerator, incubator and environmental chamber
Battery Type1 Silver Oxide battery
Accuracy±0.5°C between 0°C and 50°C, otherwise ±1°C
DisplayDigital
Width (Metric)12 cm
Unit SizeEach