Traceable™ Flashing LED Alert Big-Digit Alarm Timer
Traceable™ Flashing LED Alert Big-Digit Alarm Timer
Actual product may vary
Traceable™ Flashing LED Alert Big-Digit Alarm Timer
Unique green light flashes every 10 seconds while in countdown mode; red light flashes when zero has been reached when the alarm is sounding
Have Questions?
Catalog number CTL06-664-251
Price (SGD)
94.00
EA
Add to cart
Price (SGD)
94.00
EA
Add to cart
Fisher Scientific™ Traceable™ Flashing LED Alert Big-Digit Alarm Timer offers a massive, 1-inch-high display that permits viewing from across the lab. Unique green light flashes every 10 seconds while in countdown mode; red light flashes when zero has been reached when the alarm is sounding.
  • Flashing LED
  • Programmable memory—up to 3 memories can be stored at any time
  • Adjustable alarm volume
  • Use any countdown time from 99 miuntes, 59 seconds to 1 second
  • When zero is reached, the timer starts counting up to show time elapsed since alarming
Specifications
Product TypeHandheld/Benchtop Timer
Time Range99 min. 59 sec.
AlarmsVisual adjustable alarm volume
Accuracy0.0001
Resolution1 sec.
DisplayLED
Dimensions (L x W x H)3.5 x 2.25 x 0.63 in. (8.9 x 5.71 x 1.6 cm)
Length (English)3.5 in.
Length (Metric)8.9 cm
Width (English)2.25 in.
Width (Metric)5.71 cm
Height (English)0.63 in.
Height (Metric)1.6 cm
Weight (English)2.5 oz.
Weight (Metric)71 g
IncludesTwo AAA Alkaline batteries, traceable certificate
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEA