Traceable™ Manometer Pressure/Vacuum Gauges
Actual product may vary
Traceable™ Manometer Pressure/Vacuum Gauges
Portable units measure and display differential pressure/vacuum in 11 different units (psi, mbar etc)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPressure RangeProduct TypeWeight (Metric)
CTL06-664-19-15 to 15 psiNon-Printing Model0.14 kg
CTL15-078-190-5 to 5 psiPrinting Model0.34 kg
CTL15-078-193-100 to 100 psiPrinting Model0.34 kg
CTL06-664-21-30 to 30 psiNon-Printing Model0.14 kg
CTL06-664-22-100 to 100 psiNon-Printing Model0.14 kg
Catalog number CTL06-664-19
Price (SGD)
797.00
Each
Pressure Range:
-15 to 15 psi
Product Type:
Non-Printing Model
Weight (Metric):
0.14 kg
Price (SGD)
797.00
Each
Portable units measure and display differential pressure/vacuum in 11 different units (psi, mbar etc)
  • Portable unit measures and displays differential pressure/vacuum in: psi, mbar, bar, cm/H2O, inch/H2O, feet/H2O, mm/Hg, inch/Hg, kg/ cm2, oz./inch2 and kPa
  • Response time: 0.5 sec.
  • Accuracy: ±0.3% of full-scale between 0 and 50°C
  • Readings are shown on backlit display with 0.625 in.H (1.6 cm) digits visible from 6 ft. Feature DATA HOLD button to freeze display and capture readings
  • Memory key recalls highest and lowest readings
  • Differential feature calculates the difference between two readings
  • Zeroing is achieved at the touch of a button
  • Display times up to 99 hours, 59 minutes, 59 seconds which allows the user to record when maximum or minimum pressure is achieved
Printing Model
  • Captures and stores up to 99 readings at the press of a button
  • Datalogging permits capturing and storing up to 12,000 readings at intervals as short as 1 second or as long as 2 hours; prints sample number, current reading, date and time
  • Manual memories and datalogger memories print without the need for a computer
  • Computer output allows manual memories, datalogger memories and current programmable capturing to be streamed to a PC computer
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Dimensions (L x W x H)7.12 x 2.75 x 1.12 in. (18.1 x 7 x 2.9 cm)
For Use With (Application)Portable units measure and display differential pressure/vacuum in 11 different units (psi, mbar etc)
Height (English)1.12 in.
Height (Metric)2.9 cm
IncludesTubing, 9V battery, ABS Plastic carrying case and RS232 port
Length (English)7.12 in.
Length (Metric)18.1 cm
MaterialABS Plastic
Operating Temperature (English)32°F to 122°F
Operating Temperature (Metric)0°C to 50°C
Response Time0.5 sec.
Time Range99 Hours 59 min. 59 sec.
Weight (English)5 oz.
Weight (Metric)0.14 kg
Width (English)2.75 in.
Width (Metric)7 cm
Accuracy±0.3%
Display TypeLCD
Pressure Range-15 to 15 psi
Product TypeNon-Printing Model
Unit SizeEach