Specifications
Accuracy (Foot Candle)±4.5% of full scale reading
Accuracy (Lux)±4.5% of full scale reading
Certifications/ComplianceWith individually serial-numbered ISO 17025 certificate indicating instrument traceability to standards provided by NIST™.
Height (Metric)3.2 cm
Includes2 in. Diameter (5.1cm) photocell, 40 in. (1m) cable, 9V alkaline battery, RS-232-C serial output for connection to IBM™-compatible computer or datalogger.
Length (English)7 in.
Length (Metric)17.78 cm
ModelDual Display
Range (Foot Candle)0 to 199.9, 200 to 1999 and 2000 to 5000
Range (Lux)0 to 1999, 2000 to 19,990 and 20,000 to 50,000
Resolution (Foot Candle)0.1 to 10 ft.-c
Resolution (Lux)1 to 100 lux
Width (English)3 in.
Width (Metric)7.62 cm
DisplayLCD
TypeLight Meter
Unit SizeEach