Traceable™ Dual-Range Light Meter
Actual product may vary
Traceable™ Dual-Range Light Meter
Measures both lux and foot-candles at a rapid sampling rate of 0.4 seconds. Display update rate is adjustable to 1 or 2 seconds.
Have Questions?
Catalog NumberDisplay
CTL06-662-63LCD
Catalog number CTL06-662-63
Price (SGD)
520.00
Each
Display:
LCD
Price (SGD)
520.00
Each
Measures both lux and foot-candles at a rapid sampling rate of 0.4 seconds. Display update rate is adjustable to 1 or 2 seconds.
  • Lux measurements: three ranges from 0 to 1999, 2000 to 19,990 and 20,000 to 50,000 lux; resolution from 1 to 100 lux; accuracy of ±5% plus two digits
  • Foot-candle measurements: three ranges from 0 to 199.9; 200 to 1999 and 2000 to 5000 ft-c.; resolution from 0.1 to 10 ft-c; accuracy of ±5% plus two digits
  • Push-button access to highest, lowest and average of recorded readings
  • DATA HOLD function 'freezes' display while analog output allows readings to be captured by a recorder
  • Photodiode and color-correction filter provide cosine- and color-corrected measurements
Specifications
Accuracy (Foot Candle)±5% plus two digits
Accuracy (Lux)±5% plus two digits
Certifications/ComplianceIndividually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Height (Metric)3.8 cm
Includes2 in. Diameter (5.1cm) photocell, 40 in. (1m) cable, 9V alkaline battery and recorder output
Length (English)6.5 in.
Length (Metric)16.5 cm
ModelDual Range
Range (Foot Candle)0 to 199.9; 200 to 1999 and 2000 to 5000 ft-c
Range (Lux)0 to 1999, 2000 to 19,990 and 20,000 to 50,000 lux
Resolution (Foot Candle)0.1 to 10 ft-c
Resolution (Lux)1 to 100 lux
Width (English)3 in.
Width (Metric)7.62 cm
DisplayLCD
TypeLight Meter
Unit SizeEach