Traceable™ Benchtop Timer: 100-Minute
Traceable™ Benchtop Timer: 100-Minute
Actual product may vary
Traceable™ Benchtop Timer: 100-Minute
One timing channel and single-line display
Have Questions?
Catalog number CTL06-662-51
Price (SGD)
58.00
EA
Add to cart
Price (SGD)
58.00
EA
Add to cart
Includes one timing channel and a single-line display. Fisher Scientific™ Traceable™ Benchtop Timer: 100-Minute is complete with an LCD screen that is readable from 20 feet.
  • Memory recall
  • Timer range: 99 minutes, 59 seconds
  • Resolution: 1 sec.
  • Accuracy: 0.01%
  • Display: 0.75 in.H LCD, readable from 20 ft.
  • Alarm: 30 seconds, silenced automatically or manually
  • Dimensions: 1.3L x 6.4W x 6.4cmH (0.5 x 2.5 x 2.5 in.H)
Specifications
Product TypeHandheld/Benchtop Timer
Time Range99 min. 59 sec.
AlarmsAudible
Accuracy0.0001
Resolution1 sec.
DisplayDigital
Dimensions (L x W x H)0.5 x 2.5 x 2.5 in. (1.27 x 6.35 x 6.35 cm)
Length (English)0.5 in.
Length (Metric)1.27 cm
Width (English)2.5 in.
Width (Metric)6.35 cm
Height (English)2.5 in.
Height (English) Display0.75 in.
Height (Metric)6.35 cm
Height (Metric) Display1.91 cm
Weight (English)0.13 lb.
Weight (Metric)0.06 kg
IncludesEasel stand, clip, and magnetic back; 2-year AAA battery
For Use With (Application)For repetitive tests
Certifications/ComplianceNIST
Unit SizeEA