Wash Bottles with Adjustable Sprayer
Wash Bottles with Adjustable Sprayer
Actual product may vary
Wash Bottles with Adjustable Sprayer
Wash Bottles with Adjustable Sprayer handles hot/cold, aqueous/solvent-based solutions.
Have Questions?
Catalog NumberCapacity (Metric)
CTL03-4051000 mL
Catalog number CTL03-405
Price (SGD)
113.00
EA
Add to cart
Capacity (Metric):
1000 mL
Price (SGD)
113.00
EA
Add to cart
Wash Bottles with Adjustable Sprayer handles hot/cold, aqueous/solvent-based solutions.
  • Bottle is made of high-density polyethylene
  • Sprayer is polypropylene
  • Trigger allows one-handed, controlled delivery with light pressure
  • Engraved markings show how much fluid is left
  • Squat shape takes up little bench space and ensures a low center of gravity
Specifications
Capacity (Metric)1000 mL
IncludesPolypropylene Adjustable Sprayer
MaterialHDPE
SterilityNon-sterile
ShapeSquat
Unit SizeEA